Learn More
Medchemexpress LLC 4-Methyl-2-oxopentanoic acid | 816-66-0 | MFCD00066204 | 95.7% | 130.14 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW012722R100MG
Encompass_Preferred
4-Methyl-2-oxopentanoic acid (Standard) is the analytical standard of 4-Methyl-2-oxopentanoic acid. It is intended for research and analytical applications. 4-Methyl-2-oxopentanoic acid (α-Ketoisocaproic acid), an abnormal metabolite, is both a neurotoxin and a metabotoxin. This product is for research use only and not sold to patients.
  • Analytical standard for research and analytical applications
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC, and MS
  • Abnormal metabolite, neurotoxin, and metabotoxin
Catalog No. 50-004-54867