Learn More
Medchemexpress LLC 4-Methyl-2-oxopentanoic acid (Standard) | 816-66-0 | 95.7% | 130.14 | 10 MG

Supplier: Medchemexpress LLC HYW012722R10MG
4-Methyl-2-oxopentanoic acid (Standard) is the analytical standard of 4-Methyl-2-oxopentanoic acid. This product is intended for research and analytical applications. 4-Methyl-2-oxopentanoic acid (α-Ketoisocaproic acid), an abnormal metabolite, is both a neurotoxin and a metabotoxin.
- Used as an analytical standard and reference standard for qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS.
- Appearance: liquid (density: 1.055 g/cm3).
- Color: colorless to light yellow.
- Structure classification: ketones, aldehydes, acids.
- Initial source: endogenous metabolite, disease markers, immune system disorder, human gut microbiota metabolites, microorganisms.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.