Learn More
Medchemexpress LLC 4-Methyl-2-oxopentanoic acid (Standard) | 816-66-0 | 95.7% | 130.14 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW012722R
Encompass_Preferred
4-Methyl-2-oxopentanoic acid (Standard) is the analytical standard of 4-Methyl-2-oxopentanoic acid. Intended for research and analytical applications, it is also known as α-Ketoisocaproic acid, an abnormal metabolite, neurotoxin, and metabotoxin. This product is for research use only and not sold to patients.
  • Analytical standard for 4-Methyl-2-oxopentanoic acid
  • Suitable for research and analytical applications
  • Functions as a neurotoxin and metabotoxin
  • Used in HPLC, GC, and MS experiments
  • Appears as a colorless to light yellow liquid (Density: 1.055 g/cm3)
Catalog No. 50-004-53184