Learn More
Medchemexpress LLC 4-Methyl-2-oxopentanoic acid (standard) | 816-66-0 | 95.7% | 130.14 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW012722R50MG
Encompass_Preferred
4-Methyl-2-oxopentanoic acid (Standard) is the analytical standard of 4-Methyl-2-oxopentanoic acid. It is intended for research and analytical applications. This compound (α-Ketoisocaproic acid), an abnormal metabolite, is both a neurotoxin and a metabotoxin. The compound is the grade of analytical standard, which is the reference standard supplied assay. It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Analytical standard for research and analytical applications
  • Abnormal metabolite and neurotoxin
  • Metabotoxin
  • Suitable for HPLC, GC, and MS research experiments
Catalog No. 50-004-54917