Learn More
Medchemexpress LLC 4-methyl-3-nitrobenzoic acid | 96-98-0 | MFCD00007174 | 100.0% | 181.15 g/mol | C8H7NO4 | 500 G

Supplier: Medchemexpress LLC HYW021293500G
4-Methyl-3-nitrobenzoic acid (CAS 96-98-0) is an organic carboxylic acid used as a building block and reagent in organic synthesis and life-science research. Provided in characterized, high-purity form for laboratory use, it supports synthetic transformations, derivatization reactions, and analytical reference applications.
- High purity suitable for research applications.
- Molecular weight 181.15 g/mol.
- Chemical formula C8H7NO4.
- Available in multiple pack sizes, including 500 g.
- Certificate of analysis and safety data sheet available.
- Suitable as a building block for synthesis and analytical standards.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.