Learn More
Medchemexpress LLC 4-methyl-5-[2-(4-morpholin-4-ylanilino)pyrimidin-4-yl]-1,3-thiazol-2-amine | 693228-63-6 | MFCD14155805 | 99.5% | 368.46 g/mol | C18H20N6OS | 5 MG
SDP
Supplier:  Medchemexpress LLC HY105585MG
Encompass_Preferred
CYC-116 is a small-molecule research compound that selectively inhibits Aurora kinases A and B. It is used to probe cell cycle regulation and anticancer mechanisms in biochemical and cellular assays, and is supplied in solid and solution formats for laboratory use.
  • Potent inhibition of Aurora A and B (Ki ≈ 8 nM and 9.2 nM).
  • Suitable for biochemical and cellular assays.
  • Available as solid and as DMSO solution for convenient dosing.
  • Molecular weight 368.46 g/mol; molecular formula C18H20N6OS.
  • High reported purity (≈99.5%) for research applications.
Catalog No. 50-002-40040