Learn More
Medchemexpress LLC 4'-Methylacetophenone | 122-00-9 | 99.2% | 134.18 | 1 KG
SDP
Supplier:  Medchemexpress LLC HYW0126531KG
Encompass_Preferred
4'-Methylacetophenone is a phenolic compound utilized as a fragrance material. It occurs naturally in volatile compounds found in food and certain natural complex substances (NCS). This compound can be used in studies of the Bayer-Villiger reaction and for synthesizing functionalized terpolymers.
  • Used for blossom notes in mimosa and in the production of hawthorn type perfumes, particularly in soap perfumes.
  • Finds application in the synthesis of functionalized terpolymers for electronic devices and biological materials.
  • Utilized in studies of the Bayer-Villiger reaction, which converts ketones to esters or lactones for the synthesis of dyes and pharmaceutical intermediates.
Catalog No. 50-004-52929