Learn More
Cayman Chemical 4-MethylamInontIpyrInhydr 10mg
SDP
Supplier:  Cayman Chemical 3161210 MG
Encompass_Preferred
A major active metabolite of metamizole; inhibits A23187-induced PGE2 production in isolated mouse peritoneal macrophages at 0.1 mM; increases the paw withdrawal threshold in a rat model of carrageenan-induced hyperalgesia at 160 UG/paw; reduces LPS-induced pyrexia in rats at 60, 90, and 120 mg/kg; a potential impurity found in commercial preparations of metamizole
Catalog No. 50-297-8914