Learn More
Medchemexpress LLC 4-Methylanisole | 104-93-8 | 99.9% | 122.17 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW012835100G
Encompass_Preferred
4-Methylanisole is a food flavoring agent naturally found in Ylang Ylang fragrance oil. It is also used in air care products, cosmetics, and toys. Toxicity studies show it reduces spleen and thymus weights in adults and can cause pup mortality and reduced pup weight, both at a NOAEL of 100 mg/kg bw/day.
  • Food flavoring agent
  • Component of Ylang Ylang fragrance oil
  • Utilized in air care products
  • Used in cosmetics
  • Applied in toys
Catalog No. 50-004-55123