Learn More
Medchemexpress LLC 4-Methylanisole | 104-93-8 | 99.9% | 122.17 | 50 G

Supplier: Medchemexpress LLC HYW01283550G
4-Methylanisole (4-Methoxytoluene) is utilized as a food flavoring agent. It is naturally present in Ylang Ylang fragrance oil. This product is for research purposes only and not sold to patients.
- Functions as a food flavoring agent
- Naturally found in Ylang Ylang fragrance oil
- Available in various quantities
- High purity (99.93%)
- Includes detailed documentation
- Offers solubility information and stock solution preparation guidance
- Used in air care products, cosmetics, and toys
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.