Learn More
Medchemexpress LLC 4-Methylanisole | 104-93-8 | 99.9% | 122.17 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW012835500G
Encompass_Preferred
4-Methylanisole (4-Methoxytoluene) is a food flavoring agent naturally found in Ylang Ylang fragrance oil. It is also used in air care products, cosmetics, and toys. Toxicity studies indicate that 4-Methylanisole can reduce spleen and thymus weights in adults and cause pup mortality and reduced pup weight, with a NOAEL of 100 mg/kg bw/day.
  • Food flavoring agent
  • Naturally found in Ylang Ylang fragrance oil
  • Used in air care products, cosmetics, and toys
  • Research use only
Catalog No. 50-004-54780