Learn More
Medchemexpress LLC 4-Methylanisole | 104-93-8 | 99.93% | 122.17 | 1 KG
SDP
Supplier:  Medchemexpress LLC HYW0128351KG
Encompass_Preferred
4-Methylanisole (4-Methoxytoluene) is a chemical compound that serves as an analytical standard for research and analytical applications. It is a food flavoring agent naturally found in Ylang Ylang fragrance oil and is also utilized in air care products, cosmetics, and toys.
  • Used as an analytical standard in research and analytical applications.
  • Naturally occurs in Ylang Ylang fragrance oil.
  • Employed as a food flavoring agent.
  • Has applications in air care products, cosmetics, and toys.
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-54938