Learn More
Medchemexpress LLC 4-methylbenzothioamide | 2362-62-1 | MFCD00173750 | 99.9% | 151.23 g/mol | C8H9NS | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW017393
Encompass_Preferred
4-Methylbenzothioamide is an organic thioamide building block (C8H9NS, 151.23 g/mol) used in medicinal chemistry for synthesis and structural modification in drug discovery research.
  • High purity suitable for synthetic chemistry (reported 99.87%).
  • Useful precursor for thioamide, thiourea, and heterocycle formation.
  • Solid reagent compatible with standard organic solvents.
  • Available in multiple lab-scale pack sizes for research workflows.
Catalog No. 50-003-04205