Learn More
Cayman Chemical 4-MethylbenzylIden camphor 10g
SDP
Supplier:  Cayman Chemical 1384010 G
Encompass_Preferred
An ultraviolet light blocker used in cosmetics and sunscreen preparations that also has estrogenic activities, binding competitively to estrogen receptors and stimulating transactivation; changes reproductive organ and brain development in both males and females
Catalog No. 50-295-5202