Learn More
Medchemexpress LLC 4-Methylbenzylidene camphor | 36861-47-9 | 99.9% | 254.37 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY17587R
Encompass_Preferred
4-Methylbenzylidene camphor (4-MBC) is an endocrine disrupter that produces estrogen-like effects. Functioning as a ultraviolet (UV) filter, it may also impact normal placental formation during early pregnancy. This compound has been observed to influence cellular processes by activating PI3K/AKT and ERK1/2 signaling pathways, elevating intracellular ROS production, and inhibiting cell proliferation. It is intended for research use only.
  • Inhibits proliferation and reduces invasion of HTR8/SVneo cells
  • Induces apoptotic cell death of human trophoblast cells
  • Increases the proportion of cells in the SubG1 phase
  • Activates PI3K/AKT and ERK1/2 signaling pathways
  • Elevates intracellular ROS production
  • Impacts uterine gene expression in neonates
Catalog No. 50-003-83958