Learn More
Medchemexpress LLC 4-Methylimidazole-d6 | 1219804-79-1 | 98.3% | 88.14 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW012814S250MG
Encompass_Preferred
4-Methylimidazole-d6 is the deuterium labeled 4-Methylimidazole, intended for research use only. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements incorporated into drug molecules can be used as tracers for quantitation during the drug development process.
Catalog No. 50-004-54859