Learn More
Medchemexpress LLC 4-Methylsalicylic acid | 50-85-1 | 100.0% | C8H8O3 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY78144
Encompass_Preferred
4-Methylsalicylic acid is a methyl derivative of Salicylic acid (HY-B0167). Its derivatives inhibit the activity of alkaline phosphatases such as TNAP and IAP, and exhibit immunosuppressive activity.
  • Methyl derivative of Salicylic acid.
  • Inhibits activity of alkaline phosphatases such as TNAP and IAP.
  • Exhibits immunosuppressive activity.
Catalog No. 50-003-59143