Learn More
Medchemexpress LLC 4-Methylsalicylic acid | 50-85-1 | 99.97% | C8H8O3 | 50 G
SDP
Supplier:  Medchemexpress LLC HY7814450G
Encompass_Preferred
4-Methylsalicylic acid is a methyl derivative of Salicylic acid. This compound's derivatives are known to inhibit the activity of alkaline phosphatases, specifically tissue-nonspecific alkaline phosphatase (TNAP) and intestinal alkaline phosphatase (IAP). Additionally, these derivatives demonstrate immunosuppressive activity.
  • Derivative of salicylic acid
  • Inhibits alkaline phosphatases
  • Exhibits immunosuppressive activity
Catalog No. 50-004-33245