Learn More
Medchemexpress LLC 4-Methylsalicylic acid | 50-85-1 | MFCD00002454 | 100.0% | C8H8O3 | 1 ML

Supplier: Medchemexpress LLC HY7814410MM/1ML
4-Methylsalicylic acid is a methyl derivative of Salicylic acid that inhibits the activity of alkaline phosphatases such as TNAP and IAP. It also exhibits immunosuppressive activity, making it suitable for various research applications.
- Methyl derivative of salicylic acid
- Inhibits alkaline phosphatases (TNAP and IAP)
- Exhibits immunosuppressive activity
- Purity 99.97%
- Molecular weight 152.15
- Chemical formula C8H8O3
- Supplied as a 1 mL solution
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.