Learn More
Selleck Chemical LLC 4-Methylsalicylic acid
SDP
Supplier:  Selleck Chemical LLC S551925MG
Encompass_Preferred
4-Methylsalicylic acid (m-Cresotic acid 2-Hydroxy-4-methylbenzoic acid m-Cresotinic acid 2-Hydroxy-p-toluic acid) is used to inhibit the medium chain acyl-CoA synthetase
Catalog No. 50-000-03525