Learn More
Alkali Scientific 4 Methylumbelliferyl 2 Acetamido 2 Deoxy 6 Sulfate β D Glucopyranoside Sodium Salt, 25 Milligrams
SDP
Supplier:  Alkali Scientific LUM001CP
Encompass_Preferred
Methylumbelliferyl 2-Acetamido-2-Deoxy-6-Sulfate β-D-Glucopyranoside Sodium Salt (25 mg) is a fluorogenic substrate used to detect enzymatic activity, particularly for sulfatases. This reagent is widely employed in assays where the detection of sulfate groups in glycosaminoglycans or other sulfated polysaccharides is required. Upon enzyme cleavage, the substrate generates a fluorescent signal, providing a highly sensitive method for studying sulfatase activity in biochemical and cell-based assays. The 25 mg quantity is ideal for small-scale experiments and allows for a range of biochemical analyses in research laboratories.
Catalog No. 50-006-60107