Learn More
Sigma Aldrich Fine Chemicals Biosciences 4-Methylumbelliferyl-beta-D-glucuronide hydrate >=98% (HPLC), BioReagent, for identification of transformed plants | 881005-91-0 | MFCD09039280 | 25MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences M913025MG
Encompass
4-Methylumbelliferyl-beta-D-glucuronide hydrate >=98% (HPLC), BioReagent, for identification of transformed plants | Purity: >=98% (HPLC) | 881005-91-0 | MFCD09039280 | 25MG
Catalog No. 50-174-5080