Learn More
Bioworld 4-Methylumbelliferyl-B-D-Galactopyranoside, 1 g
Supplier:  Bioworld 215300171
Encompass_Preferred
4-Methylumbelliferyl-β-D-galactopyranosideC16H18O8MW 338.32 Fluorescent substrate for β-D-Galactosidase.Purity: >98%. 4-Methylumbelliferone <50ppm.Soluble in 1% DMF.CAS 6160-78-7. This product is sold for laboratory research use only.
Catalog No. 50-198-789