Learn More
Alkali Scientific 4 Methylumbelliferyl β D Xylopyranoside, 250 Mg
SDP
Supplier:  Alkali Scientific BDX001CP
Encompass
4 Methylumbelliferyl β D Xylopyranoside is a high-purity reagent used to detect xylanase activity in various biochemical and microbiological assays. When hydrolyzed by xylanase enzymes, it releases a fluorescent compound that is easily measured. This makes it an essential tool for studying hemicellulose breakdown and other aspects of carbohydrate metabolism in microorganisms. The 250 mg pack provides enough material for multiple experiments, offering significant value for laboratories conducting high-throughput enzymatic assays. It is particularly useful in research focused on industrial enzyme applications and the degradation of plant biomass.
Catalog No. 50-006-60678