Learn More
Medchemexpress LLC 4-Morpholinoaniline | 2524-67-6 | 99.3% | C10H14N2O | 25 G

Supplier: Medchemexpress LLC 25GHY75777
4-Morpholinoaniline is a biochemical reagent suitable for use as a biological material or organic compound in life science related research. It is intended for research use only.
- Suitable for life science related research
- Can be used as a biological material
- Functions as an organic compound
- Purity: 99.3%
- Appearance: solid, brown to black
- Soluble in DMSO (≥ 100 mg/mL)
- Store at 4°C, protected from light
- In solvent, store at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.