Learn More
Carbosynth LLC. 4-Methylumbelliferyl-alpha-D-glucopyranoside, 17833-43-1, MFCD00067661, 1 g
SDP
Supplier:  Carbosynth LLC. M56001G
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Formula: C16H18O8, Molecular weight: 338.32 g/mol, 99%, Store at -15C, Abbreviations: 4-MU-alpha-D-Glc.
Catalog No. 50-852-476