Learn More
Bioworld 4-MUG, Trihydrate, 1 g
Supplier:  Bioworld 303500002
Encompass_Preferred
4-MUG (4-Methylumbelliferyl-b-D-glucuronide)A fluorescent substrate for b-D-Glucuronidase (GUS) encoded by the gusA gene isolated originally from E.coli. Cleavage of the substrate 4-MUG by a b-glucuronidase activity leads to the generation of the fluorigenic product 4-MU, which can be visualized or detected by irradiation with UV light. The fluorescence assay allows quantitation of GUS activity by means of a fluorimeter in protein extracts in conjunction with 4-MUG at a peak excitation of 365nm (UV) and a peak emission of 455nm (blue).Technical Information:Synonym: 4-Methylumbelliferyl-b-D-glucuronide, trihydrate(4-MUG) Appearance: white powderSolubility: 50mg/mL pyridine clear, colorless Soluble in DMFO and DMSO
Catalog No. 50-254-009