Learn More
Medchemexpress LLC 4-munana | 76204-02-9 | 98.1% | 489.41 g/mol | C21H24NNaO11 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW0198231MG
Encompass_Preferred
4-MUNANA is a fluorogenic substrate for influenza virus neuraminidase (NA). Upon NA-catalyzed hydrolysis it releases fluorescent 4-methylumbelliferone (4-MU), enabling sensitive detection and quantification of neuraminidase activity in biochemical and virological assays. It is intended for research use in inhibitor screening, antiviral drug development, and mechanistic studies.
  • Fluorogenic substrate producing 4-methylumbelliferone for fluorescence detection.
  • Enables quantitative neuraminidase activity assays (Ex 317 nm / Em 410 nm).
  • Suitable for screening neuraminidase inhibitors and antiviral research.
  • High purity appropriate for biochemical assays (purity 98.08%).
  • Offered in multiple small pack sizes to support screening and method development.
Catalog No. 50-003-01852