Learn More
Medchemexpress LLC 4-N-[4-(2-methyl-3-propan-2-ylindazol-5-yl)pyrimidin-2-yl]-1-N-(oxan-4-yl)cyclohexane-1,4-diamine | 1619903-54-6 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY12293100MG
Encompass_Preferred
LY2857785 is a type I reversible, ATP-competitive kinase inhibitor that potently inhibits CDK9 and also targets transcriptional kinases CDK8 and CDK7. It is supplied as a high-purity research compound for in vitro studies of transcriptional regulation and CDK9-dependent signaling.
  • Potent CDK9 inhibitor with IC50 ≈ 11 nM.
  • Also inhibits CDK8 (IC50 ≈ 16 nM) and CDK7 (IC50 ≈ 246 nM).
  • High purity suitable for biochemical and cell-based assays.
  • Supplied as a solid powder with molecular weight 448.60 and formula C26H36N6O.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-33734