Learn More
Medchemexpress LLC 4-N,6-N-bis[(4-fluoro-3-methylphenyl)methyl]pyrimidine-4,6-dicarboxamide | 544678-85-5 | MFCD09878513 | 99.2% | 410.42 g/mol | C22H20F2N4O2 | 1MG
SDP
Supplier:  Medchemexpress LLC HY1251661MG
Encompass_Preferred
DB04760 is a potent, highly selective, non-zinc-chelating inhibitor of matrix metalloproteinase-13 (MMP-13) with a reported biochemical IC50 of 8 nM. Preclinical studies show the compound reduces paclitaxel-induced neurotoxicity and exhibits anticancer activity. The material is supplied as a light yellow to yellow solid with high purity and recommended storage conditions for powder and solution forms.
Catalog No. 50-002-38385