Learn More
Medchemexpress LLC 4-n-Octyl-anisole-d17 | 1219794-56-5 | C15H7D17O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY151186S5MG
Encompass_Preferred
4-n-Octyl-anisole-d17 is the deuterium-labeled version of 4-n-Octyl-anisole. Stable heavy isotopes, such as hydrogen and carbon, are primarily incorporated into drug molecules as tracers for quantification during drug development. Deuteration is gaining attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS
Catalog No. 50-003-74782