Learn More
Medchemexpress LLC (4-NH2)-Exatecan | 2495742-21-5 | 99.9% | C23H21N3O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14539750MG
Encompass_Preferred
(4-NH2)-Exatecan is a topoisomerase inhibitor and a derivative of Exatecan. It can be utilized in the synthesis of antibody-drug conjugates (ADCs). It contains a linker for connecting to a Ligand Unit, where the linker is attached in a cleavable manner to the amino residue, characterized by the addition of an amino (NH2) functional group at the 4th position of the parent molecule.
  • A topoisomerase inhibitor
  • Derivative of Exatecan
  • Can be utilized in the synthesis of antibody-drug conjugates (ADCs)
  • Contains a linker for connecting to a Ligand Unit
  • Linker is attached in a cleavable manner to the amino residue
  • Characterized by the addition of an amino (NH2) functional group at the 4th position
Catalog No. 50-003-55198