Learn More
Medchemexpress LLC 4-nitro-1-(triphenylmethyl)-1H-imidazole | 173865-33-3 | 98.0% | 355.39 g/mol | C22H17N3O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10090110MM 1ML
Encompass_Preferred
CDD3505 is a small-molecule research compound (4-nitro-1-(triphenylmethyl)-1H-imidazole) reported to elevate high-density lipoprotein (HDL) cholesterol by inducing hepatic cytochrome P450 3A (CYP3A) activity. It is used as a tool compound in studies of lipid metabolism and drug-enzyme interactions and is supplied in solution and solid forms for in vitro research under appropriate safety conditions.
  • Induces hepatic CYP3A enzymes and elevates HDL cholesterol.
  • Suitable for in vitro studies of lipid metabolism and drug metabolism.
  • Supplied as 10 mM solution in DMSO and as solid quantities for formulation flexibility.
  • Typical purity ≥98.0%.
  • CAS number 173865-33-3; molecular weight 355.39 g/mol.
  • Handle according to SDS and use appropriate laboratory safety practices.
Catalog No. 50-002-12254