Learn More
Medchemexpress LLC 4-nitro-benzenemethanol, 1-methanesulfonate | 39628-94-9 | 98.8% | 231.23 g/mol | C8H9NO5S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11838010MG
Encompass_Preferred
p-Nitrobenzyl mesylate (PNBM) is a thiol- and phosphorothioate-specific alkylating reagent used to modify phosphorothioates to generate epitopes that are recognized by specific antibodies. It is commonly used for kinase substrate identification and thiophosphate labeling in biochemical assays, and is supplied as a small-scale solid for laboratory use.
  • Alkylates phosphorothioates to form antibody-recognizable epitopes.
  • Applicable to direct kinase substrate identification and thiophosphate labeling.
  • High purity suitable for analytical and preparative experiments.
  • Supplied as a 10 mg solid for small-scale laboratory workflows.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-27431