Learn More
Medchemexpress LLC 4'-Nitroacetophenone | 100-19-6 | MFCD00007355 | 99.7% | 165.15 g/mol | C8H7NO3 | 500g
SDP
Supplier:  Medchemexpress LLC HYY0185500G
Encompass_Preferred
4 -Nitroacetophenone is a nitroaromatic compound 4 -Nitroacetophenone is an important organic synthetic intermediate which can be used for the synthesis of other active compounds such as Chloramphenicol (HY-B0239)[1][2][3]
Catalog No. 50-002-70057