Learn More
Medchemexpress LLC 4-nitrobenzoic acid | 62-23-7 | MFCD00007352 | 99.8% | 167.12 | C7H5NO4 | 50mg
SDP
Supplier:  Medchemexpress LLC HYY060750MG
Encompass_Preferred
4-Nitrobenzoic acid is a nitroaromatic compound that can be used in the synthesis of other active compounds 4-Nitrobenzoic acid is also an inhibitor agent which can be used for the recognition of the Mycobacterium tuberculosis complex and differentiation from non-tuberculous mycobacteria[1]
Catalog No. 50-002-68897