Learn More
Medchemexpress LLC 4-nitrobenzyl alcohol | 619-73-8 | MFCD00007376 | 98.3% | 153.14 g/mol | C7H7NO3 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW015570100G
Encompass_Preferred
4-nitrobenzyl alcohol (CAS 619-73-8) is an organic nitro-substituted benzyl alcohol used as a building block and intermediate in pharmaceutical and chemical research. It is supplied as a white to off-white solid, with molecular weight 153.14 g/mol, documented analytical data, and intended for research use only.
  • High purity suitable for synthetic applications
  • White to off-white solid form for easy handling
  • Documented analytical data including COA and HNMR
  • Soluble in common organic solvents for reaction compatibility
  • Useful as a reactant and protecting-group precursor in synthesis
Catalog No. 50-003-00726