Learn More
Medchemexpress LLC 4-nitrobenzyl alcohol | 619-73-8 | MFCD00007376 | 99.4% | 153.14 g/mol | C7H7NO3 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW015570
Encompass_Preferred
4-Nitrobenzyl alcohol is an aromatic nitro-substituted benzyl alcohol used as an intermediate and building block in organic and medicinal chemistry. It is commonly employed to introduce 4-nitrobenzyl protecting groups and as a precursor for further functionalization in multi-step syntheses. The material's solid, high-purity form supports reliable reaction performance and analytical consistency.
  • High purity (99.39%) for consistent synthetic performance.
  • Solid, white to off-white physical form for easy handling and weighing.
  • Molecular formula C7H7NO3 and molecular weight 153.14 g/mol for accurate stoichiometry.
  • Suitable as a protecting group precursor and synthetic intermediate.
  • Compatible with common organic solvents and standard laboratory procedures.
Catalog No. 50-003-03086