Learn More
Medchemexpress LLC 4-nitrobenzyl alcohol | 619-73-8 | MFCD00007376 | 99.4% | 153.14 g/mol | C7H7NO3 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW015570500G
Encompass_Preferred
4-Nitrobenzyl alcohol (4-nitrobenzenemethanol) is a para-nitro substituted benzyl alcohol commonly used as an intermediate in organic synthesis and medicinal chemistry. Supplied as a solid with high reported purity, it serves as a versatile reagent for building nitro-containing aromatic derivatives and for transformations such as reductions, protections, and coupling reactions.
  • High purity suitable for research and synthesis.
  • Solid form for straightforward storage and handling.
  • Versatile building block for aromatic nitro compound synthesis.
  • Compatible with reductions, protections, and coupling reactions.
  • Widely used in medicinal chemistry and material science research.
Catalog No. 50-003-00663