Learn More
Medchemexpress LLC 4-nitrobenzyl methanesulfonate | 39628-94-9 | MFCD25963099 | 98.8% | 231.23 g/mol | C8H9NO5S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11838050MG
Encompass_Preferred
p-Nitrobenzyl mesylate is an alkylating reagent used to modify phosphorothioates and to identify direct kinase substrates. It is supplied as a solid and in solution for biochemical research applications.
  • Alkylates phosphorothioates to form detectable epitopes.
  • Applicable for direct kinase substrate identification workflows.
  • High purity with H NMR and LCMS confirmation.
  • Available in small research pack sizes and ready-to-use solution formats.
  • Molecular formula C8H9NO5S, molecular weight 231.23 g/mol.
Catalog No. 50-002-27463