Learn More
Medchemexpress LLC 4-nitrocatechol | 3316-09-4 | MFCD00007242 | 99.7% | 155.11 | C6H5NO4 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0668905G
Encompass_Preferred
4-nitrocatechol is a small-molecule lipoxygenase inhibitor supplied for research use. It has CAS number 3316-09-4, molecular formula C6H5NO4, and molecular weight 155.11. The material is supplied as a powder with reported purity of 99.73% and is available in multiple pack sizes including 5 g. Store powder at -20°C for long-term storage; in solution, store at -80°C for longer stability.
  • potent lipoxygenase inhibitor for biochemical studies.
  • high reported purity for analytical work.
  • available in convenient small-scale pack sizes for lab use.
  • stable as a dry powder under frozen storage.
  • intended for research use only; not for human consumption.
Catalog No. 50-002-10185