Learn More
Medchemexpress LLC 4-Nitrophenyl 4-guanidinobenzoate (hydrochloride) | 19135-17-2 | 99.4% | 336.73 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY137814
Encompass_Preferred
4-Nitrophenyl 4-guanidinobenzoate (NPGB) hydrochloride is a potent sperm acrosin inhibitor and trypsin substrate. It is a white to off-white solid, intended for laboratory research.
  • Potent sperm acrosin inhibitor and trypsin substrate.
  • White to off-white solid appearance.
  • Stable under recommended storage conditions.
  • For research use only.
  • Store at 4°C in sealed storage, away from moisture.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month in sealed storage, away from moisture.
Catalog No. 50-003-47807