Learn More
Alkali Scientific 4 Nitrophenyl β D xylopyranoside, 500 Mg
SDP
Supplier:  Alkali Scientific LBE001CP
Encompass
4-Nitrophenyl β-D-xylopyranoside, 500mg, is a reagent used in the study of xylanase activity and other glycosidases. This compound is a substrate for enzymes that break down complex sugars, particularly xylan, and its hydrolysis releases a p-nitrophenol group, which can be quantified spectrophotometrically. The 500mg quantity is perfect for smaller-scale experiments or routine enzyme assays. This reagent is used in a variety of applications, including carbohydrate metabolism studies, enzyme kinetics, and the development of biofuels, where xylanase activity is crucial for breaking down plant biomass.
Catalog No. 50-006-59720