Learn More
Medchemexpress LLC 4-Nitrophenyl phosph 25mg | 330-13-2 | 219.09 g/mol | C6H6NO6P | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11602225MG
Encompass_Preferred
4-Nitrophenyl phosphate (pNPP) is a chromogenic phosphatase substrate supplied as a solid. Upon enzymatic hydrolysis it produces the yellow 4-nitrophenolate ion, which can be quantified spectrophotometrically at 405 nm. Common applications include alkaline and acid phosphatase assays, ELISA signal development, and enzyme kinetics studies.
  • Colorless solid substrate that yields a yellow 4-nitrophenolate upon hydrolysis.
  • Easy spectrophotometric readout at 405 nm for quantitative assays.
  • Suitable for alkaline and acid phosphatase assays, ELISA, and enzyme kinetics.
  • High purity for reliable activity measurements.
  • Supplied in defined milligram quantities for bench-scale experiments.
Catalog No. 50-002-29748