Learn More
Alkali Scientific 4 Nitrophenyl Phosphate Disodium Salt Hexahydrate, 1 G
SDP
Supplier:  Alkali Scientific ROP003CP
Encompass_Preferred
4-Nitrophenyl Phosphate Disodium Salt Hexahydrate, 1g, is a reagent used in biochemical and enzymatic assays to study phosphatase activity. This compound serves as a substrate for phosphatases that hydrolyze phosphoric esters, releasing a p-nitrophenol group that can be measured spectrophotometrically. The disodium salt hexahydrate form improves solubility in aqueous solutions, making it versatile for various experimental conditions. It is commonly used in enzyme kinetics studies, protein assays, and diagnostic procedures that involve phosphatase activities. The 1g quantity is suitable for small-scale experimental setups, particularly in molecular biology and biochemistry research.
Catalog No. 50-006-61675