Learn More
Medchemexpress LLC 4-Nitrophenyl Sulfate (potassium) | 6217-68-1 | 99.9% | 257.26 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW009454
Encompass_Preferred
Potassium p-nitrophenyl sulfate is a chromogenic substrate used for arylsulfatase. It is released by arylsulfonase cleavage, enabling quantitative determination of arylsulfonase activity through colorimetric detection at 400 nm. This product is for research use only.
  • Chromogenic substrate for arylsulfatase
  • Released by arylsulfonase cleavage
  • Allows quantitative determination of arylsulfonase activity
  • Activity measured by colorimetric detection at 400 nm
Catalog No. 50-003-66350