Learn More
Medchemexpress LLC 4-nitroso hydrochlorothiazide-13C,15N2,d2 | 63779-86-2 | 99.3% | C613CH5D2ClN215N2O5S2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY144140S1MG
Encompass_Preferred
4-Nitroso hydrochlorothiazide-13C,15N2,d2 is a deuterium, 13C-, and 15-labeled version of 4-Nitroso hydrochlorothiazide. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules primarily for use as tracers in the drug development process for quantitation. Deuteration is notable for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • This compound serves as a tracer.
  • It can be utilized as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS.
Catalog No. 50-003-53900