Learn More
Cayman Chemical 4-Nonylphenol-2 3 5 6-d4 5mg
SDP
Supplier:  Cayman Chemical 394975 MG
Encompass_Preferred
An internal standard for the quantification of 4-nonylphenol by GC- or LC-MS
Catalog No. 50-291-9220