Learn More
Medchemexpress LLC HY-N4300 5mg Medchemexpress, 4'-O-Methylochnaflavone CAS:49619-87-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN43005MG
Encompass_Preferred
Medchemexpress, HY-N4300 5mg 4'-O-Methylochnaflavone CAS:49619-87-6 4'-O-Methylochnaflavone is a biflavonoid isolated from Lonicera japonica, suppresses mouse lymphocyte proliferation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2932