Learn More
Cayman Chemical 4-oxo-2-Nonal Alkyn 100ug
SDP
Supplier:  Cayman Chemical 17104100 UG
Encompass_Preferred
An alkyne-tagged electrophile that can be used as a probe to isolate and identify the reaction products of lipid peroxidation using click chemistry
Catalog No. 50-296-5577